Arsenic standard solution
Arsenic, As, 1000 mg/L, H₂O, 100 mL
Catalog # HACH1457142
Supplier: Hach
Jump to:
Product Details & Documents
Standard solution for accuracy checks in arsenic determinations.
Specifications
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pk:100 mL
- Element:Arsenic
- Symbol:As



