Order Entry
Belgium
ContactUsLinkComponent
Potassium iodide-iodate standard solution, 0,0125 N
Potassium iodide-iodate standard solution, 0,0125 N
Catalog # HACH1400153
Supplier:  Hach
CAS Number:  
undefined
Potassium iodide-iodate standard solution, 0,0125 N
Catalog # HACH1400153
Supplier:  Hach
CAS Number:  

Specifications

  • Pk:
    1.000 mL
  • Description:
    Potassium iodide-iodate standard solution, 0,0125 N

Specifications

About this item

Used for determination of sulphite by burette iodometric titration.