Orders LinkContactUsLinkComponent
Detergents reagent solution
Detergents reagent solution
Detergent reagent solution in marked dropping bottle (MDB), Toluidine blue-O chloroform, 100 mL
Catalog # HACH105932
Supplier:   Hach
Detergents reagent solution
Catalog # HACH105932
Supplier:  Hach
CAS Number:  
Detergents reagent solution
Detergents reagent solution
Detergent reagent solution in marked dropping bottle (MDB), Toluidine blue-O chloroform, 100 mL
CAS Number:  
Price
47,80 €
Log in for availability
1 * 100 mL
Product Details & Documents
Used for the determination of detergents (surfactants) by the crystal violet method. For use with storm water test kit.
Packaging: Reagent is packed in a marked dropping bottle.
Specifications
Specifications
  • Chemical method:
    Toluidine blue-O chloroform
  • Pk:
    100 mL
  • Description:
    Detergent reagent solution in marked dropping bottle (MDB)
Specifications