Order Entry
Belgium
ContactUsLinkComponent
Ethanol, ISO guide 34, calibration standards
Ethanol, ISO guide 34, calibration standards
Catalog # PROOCERE-040
CAS Number:  
Ethanol, ISO guide 34, calibration standards
Catalog # PROOCERE-040
CAS Number:  

Specifications

  • Pack type:
    Ampoules
  • Pk:
    10 Ampoul
  • Standards:
    Calibration standard
  • CAS No.:
    64-17-5
  • Description:
    Ethanol-10 mg/dl
  • Seq:
    01
  • Formula:
    CH₁CH₁OH
  • Description:
    Ethanol

Specifications

About this item

Suitable for the development and validation of new analytical methodologies, for the verification of existing processes and for routine use as either calibration or control materials.

Certifications: Meets the ISO/IEC 17025 and equivalent accreditation systems.