Organic reference standards, allopurinols, Ph. Eur.
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:27511-79-1
- Formula:C₈H₁₄N₈O₆S
- Description:Allopurinol impurity A, (5-amino-1H-pyrazole-4-carboxamide )
Specifications
Frequently Bought Together