Orders LinkContactUsLinkComponent
Capsules for microbalances
Capsules for microbalances
Silver capsules with capcity: 0,15 ml, height 9 mm and external Ø: 5 mm
Catalog # LUDI176981126
Supplier:   LUDI
Capsules for microbalances
Catalog # LUDI176981126
Supplier:  LUDI
Capsules for microbalances
Capsules for microbalances
Silver capsules with capcity: 0,15 ml, height 9 mm and external Ø: 5 mm
Price
144,00 €
Log in for availability
1 * 250 ST
Product Details & Documents
Made of silver.
  • Very good chemical resistance
  • Very good temperature resistance
They can be used with all commercially available analysers and microbalances.
Specifications
  • Description:
    Silver capsules with capcity: 0,15 ml, height 9 mm and external Ø: 5 mm