Orders LinkContactUsLinkComponent
Weighing papers
Weighing papers
76,2×76,2 mm
Catalog # HEATHS120116
Supplier:   HEATHROW
Weighing papers
Catalog # HEATHS120116
Supplier:  HEATHROW
Weighing papers
Weighing papers
76,2×76,2 mm
Price
67,30 €
Log in for availability
1 * 500 ST
Product Details & Documents
These weighing papers are nitrogen-free for a very smooth surface and good sample recovery.
  • Disposable
  • Strong enough to support dense material
Colour: White
Specifications
  • Seq:
    01
  • Size:
    76,2×76,2 mm