Orders LinkContactUsLinkComponent
Covernets
Covernets
Supplier:   MIELE
Covernets
Supplier:  MIELE
Certificates
Covernets
4 Options Available Below
Product Details & Documents
Covering net for optimum fixing of delicate items to be washed.
  • Stainless steel frame covered with plastic net
  • Particularly resistant and durable
Product Family Options
Product Information
  • Availability
    Price
  • MIEL10283650
    Covering net, 215×445 mm
    Covering net, 215×445 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 151,05 €
    151,05 €
    1 * 1 ST
       151,05 €
  • MIEL10283670
    Covering net, 215×206 mm
    Covering net, 215×206 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 112,10 €
    112,10 €
    1 * 1 ST
       112,10 €
  • MIEL10283680
    Replacement net for 215×445 mm
    Replacement net for 215×445 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,70 €
    62,70 €
    1 * 1 ST
       62,70 €
  • MIEL10283690
    Replacement net for 215×206 mm
    Replacement net for 215×206 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 54,15 €
    54,15 €
    1 * 1 ST
       54,15 €