ContactUsLinkComponent
Volatile oil apparatus for oils lighter than water, SP Wilmad-LabGlass
Volatile oil apparatus for oils lighter than water, SP Wilmad-LabGlass
Catalog # WILMLG-6655-100
Supplier:  SP Industries
Volatile oil apparatus for oils lighter than water, SP Wilmad-LabGlass
Catalog # WILMLG-6655-100
Supplier:  SP Industries
Specifications
  • Description:
    Volatile oil (LTW) apparatus
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
This distilling apparatus is used to determine volatile oil in drugs by distillation of a weighed quantity of drug in contact with water.
  • The Clevenger type receiver trap included in this apparatus is for use when the oils are lighter than water
  • The condenser rests on top of the receiver
  • There is a discharge stopcock below the return tube
  • Flask and receiver are connected by a standard taper 24/40 joint
The receiver, graduated to 5 ml with 0,1 ml increments, has a tube connecting the lower part below the scale with the top of the vapour tube to return excess water to the flask.
Delivery: The apparatus also includes a 300 ml round bottom flask and a finger type condenser.