Order Entry
Belgium
Orders LinkContactUsLinkComponent
Precision hotplates with separate controller

Precision hotplates with separate controller

350 °C, 2200 W, PZ 35 EB with controller 2860 EB
Catalog # GESTPZ35EB
Supplier:   GESTIGKEIT HARRY
Precision hotplates with separate controller
Catalog # GESTPZ35EB
Supplier:  GESTIGKEIT HARRY
Precision hotplates with separate controller
Precision hotplates with separate controller

350 °C, 2200 W, PZ 35 EB with controller 2860 EB

Price
2 920,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

This precision hotplate is equipped with adjustable temperature sensors (100 to 350 °C). These sensors function as an emergency control which protects the appliance and the substance being heated from dangerous overheating.

Specifications
  • Heating capacity:
    2200 W
  • Temp.:
    350 °C
  • Type:
    PZ 35 EB with controller 2860 EB
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.