Order Entry
Belgium
Orders LinkContactUsLinkComponent
Sedimentation cones, Imhoff type

Sedimentation cones, Imhoff type

Catalog # 528-1121
Supplier:   KARTELL
Sedimentation cones, Imhoff type
Catalog # 528-1121
Supplier:  KARTELL
Sedimentation cones, Imhoff type
Sedimentation cones, Imhoff type
Price
71,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Made of Acrylic, with PP screw cap.

  • Permanently moulded graduations
  • Clear
  • Easily drained via screw plug in the base
Specifications
  • Cat. No.:
    528-1121
  • Graduation:
    1 - 40 ml
  • Top int.Ø:
    125
  • Height:
    480 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.