ContactUsLinkComponent
Volumetric flask, class A, glass stopper with individual certificate, BLAUBRAND®
Volumetric flask, class A, glass stopper with individual certificate, BLAUBRAND®
  BRND937266
 :  Brand
undefined
Volumetric flask, class A, glass stopper with individual certificate, BLAUBRAND®
  BRND937266
 :  Brand
  • Capacity:
    5000 ml
  • Description:
    Volumetric Flask NS 34/35
  • Pk:
    1
Borosilicate glass 3.3, with glass stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
  • Individual certificate included