Orders LinkContactUsLinkComponent
VWR® Storage Vials
Supplier:   LA PHA PACK
VWR® Storage Vials
Supplier:  LA PHA PACK
Certificates
VWR® Storage Vials
VWR® Storage Vials
2 Options Available Below
Product Details & Documents
Screw neck vials in clear or amber glass, hydrolytic class 1, for sample storage. A broad range of closed top screw caps are available with different types of septa.
Product Family Options
Screw neck vials for storage purposes
Product Information
  • Capacity
    Ø×H
    Colour
    Cap included
    Availability
    Price
  • 8,00 ml
    16,6×61 mm
    Clear
    No
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 61,10 €
    61,10 €
    1 * 100 ST
       61,10 €
  • 12,00 ml
    18,5×66 mm
    Clear
    No
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 649,00 €
    649,00 €
    1 * 1.000 ST
       649,00 €
Closed top PP screw caps for storage vials
Product Information
  • Catalog #
    Cap colour
    Septum
    Thickness
    Description
    Hardness
    Availability
    Price
  • White
    Butyl red/PTFE grey
    1,3 mm
    Screw cap, closed top, ND20
    55° shore A
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 58,00 €
    58,00 €
    1 * 100 ST
       58,00 €
Product Information
  • Catalog #
    Cap colour
    Septum
    Thickness
    Description
    Hardness
    Availability
    Price
  • 55° shore A
    Butyl red/PTFE grey
    1,3 mm
    PP white, closed, for 548-3231
    65
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 419,00 €
    419,00 €
    1 * 1.000 ST
       419,00 €
Product Information
  • Catalog #
    Cap colour
    Septum
    Thickness
    Description
    Hardness
    Availability
    Price
  • Black
    Butyl red/PTFE grey
    1,6 mm
    Screw cap, closed top, ND15
    55° shore A
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 36,60 €
    36,60 €
    1 * 100 ST
       36,60 €
Product Information
  • Catalog #
    Cap colour
    Septum
    Thickness
    Description
    Hardness
    Availability
    Price
  • Black
    Silicone white/PTFE red
    1,3 mm
    Screw cap, closed top, ND15
    45° shore A
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 66,60 €
    66,60 €
    1 * 100 ST
       66,60 €
Product Information
  • Catalog #
    Cap colour
    Septum
    Thickness
    Description
    Hardness
    Availability
    Price
  • Black
    Natural rubber red-orange/TEF transparent
    1,3 mm
    Screw cap, closed top, ND15
    60° shore A
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 38,10 €
    38,10 €
    1 * 100 ST
       38,10 €