VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Certificates
About this item
LDPE, disposable. These unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials.
- Will not shatter or break
- Non toxic and inert to biological fluids and most acids
- Can be sealed, refrigerated and gas sterilised
- No bulb to insert or remove
- Uniform drop size
The low affinity surface reduces the loss of cells and valuable proteins due to binding. They work well whenever there is a need for quick, safe transfer of fluids.
Ordering information: To help you find the correct product and catalogue number, please have a look at the brochure VWR® for Liquid Handling, pages 12 to 13, which you can find under 'Related information' and 'Product literature'. There are images of the pipettes available next to the catalogue number and description of packaging.
Frequently Bought Together