ContactUsLinkComponent
Micro injection port adapter assembly, Upchurch Scientific®
Micro injection port adapter assembly, Upchurch Scientific®
Catalog # UPCHM-432-03
Micro injection port adapter assembly, Upchurch Scientific®
Catalog # UPCHM-432-03
Specifications
  • Description:
    Replacement tubing/fitting assembly
  • Seq:
    02
  • Pk:
    1
Specifications
About this item
This micro injection port adapter assembly can be connected to 360 µm Ext.Ø capillary tubing to introduce samples. It accepts standard 22 gauge Hamilton-style injection syringe needles. The needle butts directly against the connecting tubing during injections, therefore no additional swept volume is added to the fluid pathway by this adapter.
  • For 360 µm Ext.Ø. tubing
  • Mount on bracket or bulkhead