Orders LinkContactUsLinkComponent
BCR® flat mop systems
BCR® flat mop systems
Supplier:   BERKSHIRE
BCR® flat mop systems
Supplier:  BERKSHIRE
Certificates
BCR® flat mop systems
2 Options Available Below
Product Details & Documents
Efficient cleaning of floors, walls and other large surfaces is critical to the overall cleanliness of the production environment. BCR® Mop Systems replicate the most effective way to clean a surface – wiping. This mop system is easy to use, highly effective and quick. The system is very flexible, combining a wide range of alternative mop wipes designed for particular levels of cleanliness with a range of flat mop heads.
  • High quality anodised aluminum
  • Can be autoclaved for use in sterile applications
  • Unique stainless steel spring mechanisms for wiper attachment
  • Plastic eyes hold mop wipers in place (not autoclavable)
Certifications: ISO Class 3-7 / floors, walls and ceilings.
Product Family Options
Flat mop heads
Product Information
  • Dimensions
    Availability
    Price
  • 141-1701
    Flat mop head 3
    Flat mop head 3
    90×280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 96,20 €
    96,20 €
    1 * 1 ST
       96,20 €
Flat mop handle
Product Information
  • Dimensions
    Availability
    Price
  • BERKHANDLE.FMTELE1
    Telescopic handle
    Telescopic handle
    950 - 1800 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,20 €
    59,20 €
    1 * 1 ST
       59,20 €
Accessories
Product Information
  • Catalog #
    Colour
    L×W
    Availability
    Price
  • MicroSeal SuperSorb® wipes, ultrasonically sealed edge
    MicroSeal SuperSorb® wipes, ultrasonically sealed edge
    BERKMSSS.0909.8
    White
    230×230 mm
    Log in for availability
    1 * 800 ST - 1 850,00 €
Product Information
  • Catalog #
    Packed
    L×W
    Availability
    Price
  • SatPax® 670 with 50% IPA / 50% DIW
    SatPax® 670 with 50% IPA / 50% DIW
    141-0113
    25 per bag, 12 packs
    230×580 mm
    Log in for availability
    1 * 300 ST - 421,00 €