Orders LinkContactUsLinkComponent
FlashGel® camera
FlashGel® camera
Supplier:   LONZA
FlashGel® camera
Supplier:  LONZA
Certificates
FlashGel® camera
FlashGel® camera
2 Options Available Below
Product Details & Documents
The FlashGel® camera captures data from the FlashGel® system, allowing complete separation, recovery and documentation safely, at the bench, in minutes. This simple digital camera in an enclosed hood connects directly to a laptop, PC or PDA via USB.
  • Complete gel run and image capture in five minutes
  • Photograph gels at the bench, without UV light
  • Sharp, clear, high-resolution images
  • Simple user interface
  • Optimised exposure for FlashGel® cassettes
Hood (W×L×H): 100×110×160 mm
Image file type: .JPG, .TIFF, .PDF
Product Family Options
Product Information
  • Availability
    Price
  • 730-1346
    FlashGel® camera, includes camera, hood enclosure, USB cable and software
    FlashGel® camera, includes camera, hood enclosure, USB cable and software
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 260,00 €
    1 260,00 €
    1 * 1 ST
       1 260,00 €
  • LONZ57062
    FlashGel® device pack, includes camera, dock and a power supply
    FlashGel® device pack, includes camera, dock and a power supply
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 2 940,00 €
    2 940,00 €
    1 * 1 KIT
       2 940,00 €