ContactUsLinkComponent
Ion selective gas sensor, carbon dioxide
Ion selective gas sensor, carbon dioxide
Catalog # 662-2810
Supplier:  Mettler - Toledo
Ion selective gas sensor, carbon dioxide
Catalog # 662-2810
Supplier:  Mettler - Toledo
Specifications
  • Type:
    Gas sensing combination electrode, carbon dioxide
  • Pk:
    1
  • Temp.:
    0...+50 °C
  • pH:
    <2
Specifications
About this item
Ion selective and gas sensitive electrodes allow the quick and easy determination of concentration and activity of dissolved ions and carbon dioxide.