Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tubes
Test Tubes
Tubes, TallPrep™, empty, MP Biomedicals
Tubes, TallPrep™, empty, MP Biomedicals
Tubes, TallPrep™, empty, MP Biomedicals
Catalog # ICNA116970000
Supplier:
MP Biomedicals
Expand image
Tubes, TallPrep™, empty, MP Biomedicals
Catalog # ICNA116970000
Supplier:
MP Biomedicals
Specifications
Capacity:
4,5 ml
Description:
Empty tubes with caps
Pk:
100
Specifications
About this item
Product Details
Product Details