dNTP mix, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
This dNTP mix with dATP, dCTP, dGTP, dTTP is suitable for use in PCR, real-time PCR, high fidelity and long PCR, LAMP-PCR, cDNA synthesis, RT-PCR, DNA sequencing and labeling.
Frequently Bought Together