ContactUsLinkComponent
PCR mix, ready to use, Taq-&GO™, MP Biomedicals
PCR mix, ready to use, Taq-&GO™, MP Biomedicals
  ICNA11EPTAG100
 :  
PCR mix, ready to use, Taq-&GO™, MP Biomedicals
  ICNA11EPTAG100
 :  
  • Pk:
    1 mL
  • Description:
    PCR mix, Taq-&GO™
Taq-&GO™ is a ready to use optimised reaction mix for PCR.
  • Significant savings in preparation time
  • Reduced pipetting
  • Reduced risks of errors and contamination
  • Stable at 4 °C
Each mix is provided 5X concentrated and can be used to perform 20 μl or 50 μl reactions. PCR Mastermixes contain all the components for PCR with the exception of template and primers.