ContactUsLinkComponent
Alkaline detergent, RBS A 285 SOLID PF
Alkaline detergent, RBS A 285 SOLID PF
  CHEPRBSA285SOLIDPF
 :  CHEMICAL
undefined
Alkaline detergent, RBS A 285 SOLID PF
  CHEPRBSA285SOLIDPF
 :  CHEMICAL
  • Description:
    Alkaline detergent, RBS A 285 SOLID PF
  • Pk:
    4,5 kg
  • pH range:
    11.4 (0.3%)
High performance alkaline detergent in powder form for use in washing machine - labwashers.
  • Phosphate-free
  • High detergency power
  • Low working concentration
RBS A 285 SOLID PF is recommended for cleaning glassware and laboratory equipment in automatic washing machine - labwashers having dispensing compartment for powdered detergent. It efficiently removes organic and biological residues, distilation residues etc.
 : Available in box of 2 × 4,5 kg. Please contact Avantor for details.
 : Not recommended for use on white metals.