Order Entry
Belgium
ContactUsLinkComponent
Phosphate-Buffered saline (PBS), 10X concentrate, pH 7,4, molecular biology grade, Corning®
Phosphate-Buffered saline (PBS), 10X concentrate, pH 7,4, molecular biology grade, Corning®
Catalog # 46-013-CM
Supplier:  Corning
CAS Number:  
undefined
Phosphate-Buffered saline (PBS), 10X concentrate, pH 7,4, molecular biology grade, Corning®
Catalog # 46-013-CM
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    6
  • Description:
    10X Phosphate-Buffered Saline (PBS)
  • Pk Info:
    6× 1 L

Specifications

About this item

Phosphate Buffered Saline (PBS) is used to maintain a physiological pH and osmolarity, two key factors in tissue culture and molecular biology research

  • Manufactured in a cGMP environment
  • Formulated with high quality WFI water
  • RNase -, DNase-, and Protease-free
  • PBS without calcium and magnesium
  • pH 7.4 ±0.1

Phosphate Buffered Saline is used to maintain a physiological pH and osmolarity, two key factors in tissue culture and molecular biology research. This concentrated form should be diluted to a working concentration using reagent grade water.

Caution: For research use only.