Order Entry
Belgium
Orders LinkContactUsLinkComponent
Digital multi timer

Digital multi timer

78×35×90 mm
Catalog # AMARE920410
Supplier:   Amarell
Digital multi timer
Catalog # AMARE920410
Supplier:  Amarell
Digital multi timer

78×35×90 mm

Price
22,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Quadruple count down/count up timer with loud alarm signal.

  • Timing capacity: 99 hours, 59 minutes
  • With stand for bench top use

Delivery information: Supplied with 1,5 V AAA battery.

Specifications
  • W×D×H:
    78×35×90 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.