ContactUsLinkComponent
Guard Cartridge Holder for HPLC Columns
  80101/N
 :  Avantor
   Low Manufacturing Impact
Guard Cartridge Holder for HPLC Columns
  80101/N
 :  Avantor
   Low Manufacturing Impact
  • Description:
    All-Guard cartridge holder for 2,1 to 4,6 mm ID columns
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001:2015 (530840 UM15)
  • Pk:
    1
This guard cartridge holder promotes increased column lifetime.
  • Improved hardware
  • Higher pressure rating
  • Low internal swept volume
The hardware of the Alltech All-Guard cartridge system has been improved and all guard cartridges and holders are now supplied in this new hardware, leading to more robust and higher efficiency separations. Chromatographic selectivity will be unaffected by this change.