ContactUsLinkComponent
VWR® pHenomenal® Dissolved Oxygen Probe
VWR® pHenomenal® Dissolved Oxygen Probe
Catalog # 664-0042
Supplier:  VWR Collection
undefined
VWR® pHenomenal® Dissolved Oxygen Probe
Catalog # 664-0042
Supplier:  VWR Collection
Specifications
  • Connection:
    3 m cable, 8-pin
  • Type:
    pHenomenal OXY11-3, galvanic
  • Description:
    pHenomenal OXY11-3, galvanic (resolution 0,1 mg/L) probe
  • Material:
    Epoxy
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Delivery: Includes DO sensor including calibration vessel, cleaning solution, electrolyte, abrasive foil and manual.