Separating funnels, Squibb, HDPE stopper, PTFE stopcock
HACH52049
: Hach
- Capacity:500 ml
- Pk:1
Pear-shaped (Squibb) glass separatory funnel with PTFE stopcock and HDPE stopper.
- Clear glass body allows for easy detection of layers between immiscible liquids
- Highly chemical resistant and fully autoclavable
- HDPE stopper with closed bottom