Orders LinkContactUsLinkComponent
Racks for test tubes
Racks for test tubes
Supplier:   Hach
Racks for test tubes
Supplier:  Hach
Certificates
Racks for test tubes
Racks for test tubes
2 Options Available Below
Product Details & Documents
These test tube racks features a special blend of polyethylene which sinks in water, assuring non-floating racks when immersed.
  • Stackable
  • Good chemical resistance
  • Good temperature resistance
Product Family Options
Product Information
  • Hole Ø
    No. of Holes
    Material
    W×D×H
    Availability
    Price
  • 20 mm
    40 (4×10)
    PE
    246×107×64 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 41,80 €
    41,80 €
    1 * 1 ST
       41,80 €
  • 24 mm
    PE
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,10 €
    25,10 €
    1 * 1 ST
       25,10 €