Order Entry
Belgium
ContactUsLinkComponent
DNA isolation, Genomic DNA isolation kit, GNOME®, MP Biomedicals
DNA isolation, Genomic DNA isolation kit, GNOME®, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
DNA isolation, Genomic DNA isolation kit, GNOME®, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • Method / Format:
    Precipitation
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
    Bacteria
    Yeast/Fungi
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    GNOME® DNA Isolation Kit

Specifications

Frequently Bought Together