Order Entry
Belgium
ContactUsLinkComponent
GeneClean® PCR clean-up, turbo for PCR kit, MP Biomedicals
GeneClean® PCR clean-up, turbo for PCR kit, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
undefined
GeneClean® PCR clean-up, turbo for PCR kit, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    GeneClean® Turbo for PCR Kit

Specifications

Frequently Bought Together