Cleanroom mops, PPSC Micro TEC
141-1393
- Description:PPSC Micro TEC, 130 g
- Length:400 mm
- Pk:1
Mops provides top cleaning performance. Made up of 100% PES super microfiber with soft-brush.
- Ready-to-use pre-washed
- Backfilling with cover sheet, external trimming TEC
- Pocket of harness extremely tear-proof
Germ reduction of bacteria, yeast fungus and mould of over 99 per cent will be reached by using water (Testing method DGMH 2001. standard method 14).
: Certified according to the requirements of the Nordic Ecolabel.