Order Entry
Belgium
ContactUsLinkComponent
Xylene (mixture of isomers) ≥98.5% (by GC) for trace metal analysis, Macron Fine Chemicals™
Flammable
Irritant
Xylene (mixture of isomers) ≥98.5% (by GC) for trace metal analysis, Macron Fine Chemicals™
Catalog # 8802-04
CAS Number:  
undefined
Flammable
Irritant
Xylene (mixture of isomers) ≥98.5% (by GC) for trace metal analysis, Macron Fine Chemicals™
Catalog # 8802-04
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥98,5%
  • Assay (m,p,o-xylenes and ethylbenzene) (by GC):
    ≥ 98.5 %
  • Acidity (µeq/g):
    ≤ 0.3
  • Arsenic and Antimony (as As):
    ≤ 0.010 ppm
  • Chloride (Cl):
    ≤ 3 ppm
  • Color (APHA):
    ≤ 10
  • Heavy Metals (as Pb):
    ≤ 1 ppm
  • Identification (by IR):
    Passes Test
  • Phosphate (PO₄):
    ≤ 1 ppm
  • Residue after Evaporation:
    ≤ 5 ppm
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Sulfur Compounds (as S):
    ≤ 0.003 %
  • Water (H₂O):
    ≤ 0.02 %
  • Trace Impurities - Aluminum (Al):
    ≤ 0.100 ppm
  • Trace Impurities - Barium (Ba):
    ≤ 1.000 ppm
  • Trace Impurities - Boron (B):
    ≤ 0.100 ppm
  • Trace Impurities - Cadmium (Cd):
    ≤ 1.000 ppm
  • Trace Impurities - Calcium (Ca):
    ≤ 0.100 ppm
  • Trace Impurities - Chromium (Cr):
    ≤ 0.100 ppm
  • Trace Impurities - Cobalt (Co):
    ≤ 0.100 ppm
  • Trace Impurities - Copper (Cu):
    ≤ 0.10 ppm
  • Trace Impurities - Gallium (Ga):
    ≤ 0.500 ppm
  • Trace Impurities - Germanium (Ge):
    ≤ 1.000 ppm
  • Trace Impurities - Gold (Au):
    ≤ 0.100 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 0.1 ppm
  • Trace Impurities - Lead (Pb):
    ≤ 0.100 ppm
  • Trace Impurities - Lithium (Li):
    ≤ 1.000 ppm
  • Trace Impurities - Magnesium (Mg):
    ≤ 0.100 ppm
  • Trace Impurities - Manganese (Mn):
    ≤ 0.100 ppm
  • Trace Impurities - Nickel (Ni):
    ≤ 0.1 ppm
  • Trace Impurities - Potassium (K):
    ≤ 0.100 ppm
  • Trace Impurities - Silicon (Si):
    ≤ 1.000 ppm
  • Trace Impurities - Silver (Ag):
    ≤ 0.500 ppm
  • Trace Impurities - Sodium (Na):
    ≤ 0.100 ppm
  • Trace Impurities - Strontium (Sr):
    ≤ 1.000 ppm
  • Trace Impurities - Tin (Sn):
    ≤ 0.100 ppm
  • Trace Impurities - Zinc (Zn):
    ≤ 0.100 ppm

Specification Test Results

About this item

 :
 :106.17 g/mol
Merck Index:13,10136
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–34 °C
Density:0.86…0.88 g/cm³ (20 °C)
Flash Pt:25 °C
EINECS:215-535-7
UN:1307
ADR:3,III
 :Management of Change (MOC) category = N
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:1330-20-7
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264