Order Entry
Belgium
ContactUsLinkComponent
Xylene (mixture of isomers) ≥98.0%, AR®, Macron Fine Chemicals™
Xylene (mixture of isomers) ≥98.0%, AR®, Macron Fine Chemicals™
Catalog # 6781-25
CAS Number:  
undefined
Xylene (mixture of isomers) ≥98.0%, AR®, Macron Fine Chemicals™
Catalog # 6781-25
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (GC):
    Min. 98,0%
  • Water (KF):
    Max. 0,05%
  • Titrable acid (meq/g):
    Max. 0,0005
  • Residue after evaporation:
    Max. 0,002%
  • Substances darkened by H₂SO₄:
    Passes test
  • Sulphur compounds:
    Max. 0,001%

Specification Test Results

About this item

 :
 :106.17 g/mol
UN:1307
ADR:3,III
CAS Number:1330-20-7
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264