Orders LinkContactUsLinkComponent
SSPE Buffer, 20X concentrate, pH 7,4
SSPE Buffer, 20X concentrate, pH 7,4
SSPE buffer 20X (3,0 M sodium chloride, 0,2 M sodium hydrogen phosphate, 0,02 M EDTA, pH 7,4) DEPC treated, 1 L
Catalog # ROCKMB-044
SSPE Buffer, 20X concentrate, pH 7,4
Catalog # ROCKMB-044
CAS Number:  
SSPE Buffer, 20X concentrate, pH 7,4
SSPE buffer 20X (3,0 M sodium chloride, 0,2 M sodium hydrogen phosphate, 0,02 M EDTA, pH 7,4) DEPC treated, 1 L
CAS Number:  
Price
248,00 €
Log in for availability
1 * 1 L
Product Details & Documents
20X SSPE buffer (3,0 M sodium chloride, 0,2 M sodium hydrogen phosphate, 0,02 M EDTA, pH 7,4) - DEPC treated.
This product was aseptically filtered through a Millipore 0,22 µm filter into clean, pre-sterilised containers. The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.
Caution: This product is for research use only and is not intended for therapeutic or diagnostic applications.
Specifications
Specifications
  • Pk:
    1 L
  • Description:
    SSPE buffer 20X (3,0 M sodium chloride, 0,2 M sodium hydrogen phosphate, 0,02 M EDTA, pH 7,4) DEPC treated
Specifications
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.