Orders LinkContactUsLinkComponent
ORP test solution
ORP test solution, 250 mL
Catalog # 662-8251
Supplier:   WTW
ORP test solution
Catalog # 662-8251
Supplier:  WTW
CAS Number:  
ORP test solution
ORP test solution, 250 mL
CAS Number:  
Price
54,10 €
Log in for availability
1 * 250 mL
Product Details & Documents
ORP test solution for electrolyte cleaning agents to test, service ORP electrodes easy conditioning and cleaning.
Documents
Specifications
Specifications
  • Pk:
    250 mL
  • Description:
    ORP test solution