Orders LinkContactUsLinkComponent
Cryoboxes, PP
Cryoboxes, PP
Supplier:   RATIOLAB
Cryoboxes, PP
Supplier:  RATIOLAB
Certificates
Cryoboxes, PP
6 Options Available Below
Product Details & Documents
Boxes made of high quality PP.
  • Autoclavable
  • Temperature-resistant from –90 to +121 °C
  • Numerically coded
  • Very good chemical resistance
With an numerical coding and corresponding vents for draining condensed water. The boxes are adjustable in height.
Product Family Options
Product Information
  • Compartments
    Colour
    W×D×H ext.
    Availability
    Price
  • 7×7
    Green
    133×133×50/75 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 41,40 €
    41,40 €
    1 * 5 ST
       41,40 €
  • 7×7
    Blue
    133×133×50/75 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 41,40 €
    41,40 €
    1 * 5 ST
       41,40 €
  • 7×7
    Black
    133×133×50/75 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 41,40 €
    41,40 €
    1 * 5 ST
       41,40 €
  • 7×7
    Multicolour
    133×133×50/75 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 41,40 €
    41,40 €
    1 * 5 ST
       41,40 €
  • 6×6
    Natural
    133×133×50/75 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 41,40 €
    41,40 €
    1 * 5 ST
       41,40 €
  • 6×6
    Green
    133×133×50/75 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 41,40 €
    41,40 €
    1 * 5 ST
       41,40 €