ContactUsLinkComponent
Sample preparation kit, AdvanceBio
Sample preparation kit, AdvanceBio
Catalog # HEWL5190-8002
Supplier:  Agilent
Sample preparation kit, AdvanceBio
Catalog # HEWL5190-8002
Supplier:  Agilent
Specifications
  • Description:
    AdvanceBio n-glycan deglycosylation cleanup cartridges, 24 samples
  • Pk:
    1
Specifications
About this item
This kit consist the enzymes and cleanup cartridges for the enzymatic deglycosylation process of N-glycan.
Deglycosylation kit includes PNGase F enzyme, phosphate reaction buffer, TRIS reaction buffer, detergent solution, denaturation solution.
Labelling kit includes 75 µl 2-aminobenzeamide solution and 75 µl reductant (sodium cyanoborohydride) solution.
Delivery: Deglycosylation kit, labelling kit and cleanup cartridges.