ContactUsLinkComponent
Insulin-Transferrin-Selenium (ITS) 100x Growth Supplement, Corning®
Insulin-Transferrin-Selenium (ITS) 100x Growth Supplement, Corning®
  MDTC25-800-CR
 :  Corning
 :  
undefined
Insulin-Transferrin-Selenium (ITS) 100x Growth Supplement, Corning®
  MDTC25-800-CR
 :  Corning
 :  
  • Pack type:
    Plastic bottle
  • Pk:
    10 mL
  • Description:
    Insulin-Transferrin-Selenium (ITS) Growth Supplement, Sterile, 100X
  • Seq:
    52
Animal free formulation
Contains:
1000 mg/L insulin, human recombinant
550 mg/L transferrin, human recombinant
0.67 mg/L selenious acid
 : Storage 2°C to 8°C. For research use only.