Paramat, Solidification point: 55 - 58 °C, in pastille form, Gurr™, without DMSO
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:8002-74-2
:8012-95-1
Specification Test Results
- Cloud point (65 °C):Clear
- Cutting/ribboning:Complies
- Congealing temperature:55 - 58 °C
Specification Test Results
Related Information
Frequently Bought Together