Orders LinkContactUsLinkComponent
PIG® mini drip deck
PIG® mini drip deck
Supplier:   NEW PIG
PIG® mini drip deck
Supplier:  NEW PIG
Certificates
PIG® mini drip deck
PIG® mini drip deck
2 Options Available Below
Product Details & Documents
If containers leak, drip or crack, make sure a Mini Drip Deck with built-in sump is in position to catch the liquid.
  • Translucent sidewalls let you easily inspect fluid level in sump
  • Compact size fits on tabletop or counter; ideal for use in labs
  • Pour spout for easy draining
Chemical-resistant polyethylene tray features built-in sump containment to trap leaks and drips whilst you work.
Caution: If using this product with flammable liquids, please consider the regulations that apply to storage and handling of flammable liquids and the safety of this application, specifically flammable vapours, static discharge and heat sources.
Product Family Options
Product Information
  • Capacity
    W×D×H
    Availability
    Price
  • 121-0535
    Mini drip deck
    Mini drip deck
    5,6 L
    430×305×100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,30 €
    52,30 €
    1 * 1 ST
       52,30 €
  • 121-0536
    Mini drip deck
    Mini drip deck
    11,3 L
    530×430×100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 84,20 €
    84,20 €
    1 * 1 ST
       84,20 €