Order Entry
Belgium
ContactUsLinkComponent
Sodium chloride, anhydrous ≥99% ACS, free-flowing, Fluka™
Sodium chloride, anhydrous ≥99% ACS, free-flowing, Fluka™
Certificates
Sodium chloride, anhydrous ≥99% ACS, free-flowing, Fluka™
Certificates

Specification Test Results

  • Assay:
    99.0 - 101.5 %
  • Turbidity of solution:
    complying
  • Colour of the solution:
    complying
  • Flowability:
    complying
  • pH (5 %, 25 °C):
    5.0 - 9.0
  • Water insoluble matter:
    Max. 0.005 %
  • Barium (Ba):
    complying
  • Calcium (Ca):
    Max. 0.002 %
  • Iron (Fe):
    Max. 2 ppm
  • Potassium (K):
    Max. 0.005 %
  • Magnesium (Mg):
    Max. 0.001 %
  • Heavy metals (ICP) ACS:
    Max. 5 ppm
  • Bromide (Br):
    Max. 0.01 %
  • Chlorate, Nitrate (as NO₃):
    Max. 0.003 %
  • Iodide (I):
    Max. 0.002 %
  • Phosphate (PO₄):
    Max. 5 ppm
  • Sulfate (SO₄):
    Max. 0.004 %

Specification Test Results

Related Information

Frequently Bought Together