- Antibody type:Primary
- Antigen name:Glycosyltransferase-like Domain Containing 1
- Antigen symbol:GTDC1
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:79712
- Antigen synonyms:GTDC1_HUMAN.|Mannosyltransferase candidate|Glycosyltransferase like 1|Glycosyltransferase like domain containing 1|Glycosyltransferase like domain containing protein 1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Glycosyltransferases that mediate the regio- and stereoselective transfer of sugars, such as the fucosyltransferases, determine cell surface-carbohydrate profiles, which is an essential interface for biological recognition processes. GTDC1 (Glycosyltransferase-like domain-containing protein 1), also known as Mat-Xa, is a 458 amino acid protein belonging to the glycosyltransferase 1 family. GTDC1 is ubiquitously expressed, with highest levels found in peripheral blood leukocytes, spleen, lung and testis.An extremely rare recessive genetic disorder, Alstré°‰ syndrome, is caused by mutations in the ALMS1 gene, which maps to chromosome 2.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GTDC1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat