Order Entry
Belgium
ContactUsLinkComponent
 
Anti-RCC2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-7768R-A555
 :  Bioss
Anti-RCC2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-7768R-A555
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Regulator of Chromosome Condensation 2
  • Antigen symbol:
    RCC2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    55920
  • Antigen synonyms:
    TD60|Protein RCC2|Telophase disk protein of 60 kDa|TD-60|KIAA1470|RCC1-like protein TD-60|RCC2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at –20 °C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    455-490/522
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Required for completion of mitosis and cytokinesis. May function as a guanine nucleotide exchange factor for the small GTPase RAC1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RCC2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat