Order Entry
Belgium
ContactUsLinkComponent
 
Anti-ITIH1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-7018R-A555
 :  Bioss
Anti-ITIH1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
  BOSSBS-7018R-A555
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Inter-alpha (globulin) Inhibitor H1
  • Antigen symbol:
    ITIH1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    3697
  • Antigen synonyms:
    ITI-HC1|Inter-alpha-trypsin inhibitor heavy chain H1|IGHEP1|ITIH1|SHAP|Serum-derived hyaluronan-associated protein|ITI heavy chain H1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

May act as a carrier of hyaluronan in serum or as a binding protein between hyaluronan and other matrix protein, including those on cell surfaces in tissues to regulate the localization, synthesis and degradation of hyaluronan which are essential to cells undergoing biological processes.Contains a potential peptide which could stimulate a broad spectrum of phagocytotic cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ITIH1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat