Order Entry
Belgium
ContactUsLinkComponent
 
Anti-APOBR Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-6336R-FITC
 :  Bioss
Anti-APOBR Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-6336R-FITC
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Apolipoprotein B Receptor
  • Antigen symbol:
    APOBR
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    55911
  • Antigen synonyms:
    APOB48R|APOB100R
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

APOB48R is a macrophage receptor that binds to the apolipoprotein B48 of dietary triglyceride (TG) rich lipoproteins. It may provide essential lipids, lipid soluble vitamins and other nutrients to reticuloendothelial cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: APOBR
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat