Order Entry
Belgium
ContactUsLinkComponent
Anti-RASA3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-RASA3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-5954R-A350
Supplier:  Bioss
Anti-RASA3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-5954R-A350
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ras GTPase-activating Protein 3
  • Antigen symbol:
    RASA3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    22821
  • Antigen synonyms:
    GAPIII|GAP1IP4BP
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    510-560/834
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Inhibitory regulator of the Ras-cyclic AMP pathway. Binds inositol tetrakisphosphate (IP4) with high affinity. Might be a specific IP4 receptor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RASA3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat