Order Entry
Belgium
ContactUsLinkComponent
Anti-ADAMTSL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-ADAMTSL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-5862R-FITC
Supplier:  Bioss
Anti-ADAMTSL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-5862R-FITC
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ADAMTS-Like 2
  • Antigen symbol:
    ADAMTSL2
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    9719
  • Antigen synonyms:
    ATL2_HUMAN.|ADAMTS like protein 2|ADAMTSL 2|ADAMTSL2|ADAMTS like 2|ADAMTSL-2|ADAMTS-like protein 2
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

ADAMTS (A Disintegrin And Metalloproteinase Domain with Thrombospondin type 1 Modules) is a family of zinc-dependent proteases that are implicated in a variety of normal and pathological conditions, including arthritis and cancer. ADAMTS protein family members contain an amino-terminal propeptide domain, a metalloproteinase domain, a disintegrin-like domain and a carboxy-terminus that contains a varying number of Thrombospondin type 1 (TSP-1) motifs. ADAMTS-L2 (ADAMTS-like protein 2) is a 951 amino acid secreted protein that is highly expressed in lung, kidney and liver. Mutations in the gene encoding ADAMTS are the cause of geleophysic dysplasia, an autosomal recessive disorder characterized by cardiac vavular anomalies, short stature, thick skin and brachydactyly. In individuals affected with geleophysic dysplasia, there is a significant increase in total active TGF-beta 1 and nuclear locations of p-SAMD2 in fibroblasts. Interestingly, ADAMTS-L2 interacts with LTBP-1, a glycoprotein that is part of the platelet-derived TGF-beta 1 complex.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADAMTSL2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat