Specifications
- Antibody type:Primary
- Antigen name:G3BP Stress Granule Assembly Factor 1
- Antigen symbol:G3BP1
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Pig,Human
- Cross adsorption:No
- Form:liquid
- Gene ID:10146
- Antigen synonyms:G3BP|Ras GTPase-activating protein-binding protein 1|ATP-dependent DNA helicase VIII|HDH-VIII|MGC111040
- Modification:phospho-Ser232
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:215-260/466
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves MYC mRNA preferentially at the 3'-UTR. ATP- and magnesium-dependent helicase. Unwinds preferentially partial DNA and RNA duplexes having a 17 bp annealed portion and either a hanging 3' tail or hanging tails at both 5'- and 3'-ends. Unwinds DNA/DNA, RNA/DNA, and RNA/RNA substrates with comparable efficiency. Acts unidirectionally by moving in the 5' to 3' direction along the bound single-stranded DNA.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: G3BP1Ser232
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Pig