Order Entry
Belgium
ContactUsLinkComponent
Anti-G3BP1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-G3BP1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-5376R-A350
 :  Bioss
Anti-G3BP1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-5376R-A350
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    G3BP Stress Granule Assembly Factor 1
  • Antigen symbol:
    G3BP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Pig,
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    10146
  • Antigen synonyms:
    G3BP|Ras GTPase-activating protein-binding protein 1|ATP-dependent DNA helicase VIII|HDH-VIII|MGC111040
  • Modification:
    phospho-Ser232
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    215-260/466
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves MYC mRNA preferentially at the 3'-UTR. ATP- and magnesium-dependent helicase. Unwinds preferentially partial DNA and RNA duplexes having a 17 bp annealed portion and either a hanging 3' tail or hanging tails at both 5'- and 3'-ends. Unwinds DNA/DNA, RNA/DNA, and RNA/RNA substrates with comparable efficiency. Acts unidirectionally by moving in the 5' to 3' direction along the bound single-stranded DNA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: G3BP1Ser232
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Pig