Order Entry
Belgium
ContactUsLinkComponent
Anti-LONP1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-LONP1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-4245R-A350
Supplier:  Bioss
Anti-LONP1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-4245R-A350
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    lon peptidase 1 peroxisomal
  • Antigen symbol:
    LONP1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    9361
  • Antigen synonyms:
    CODASS|hLON|LONP|PRSS15|LON|PIM1|LonHS
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    375-420/959
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

ATP-dependent serine protease that mediates the selective degradation of misfolded, unassembled or oxidatively damaged polypeptides as well as certain short-lived regulatory proteins in the mitochondrial matrix. May also have a chaperone function in the assembly of inner membrane protein complexes. Participates in the regulation of mitochondrial gene expression and in the maintenance of the integrity of the mitochondrial genome. Binds to mitochondrial promoters and RNA in a single-stranded, site-specific, and strand-specific manner. May regulate mitochondrial DNA replication and/or gene expression using site-specific, single-stranded DNA binding to target the degradation of regulatory proteins binding to adjacent sites in mitochondrial promoters. Endogenous substrates include mitochondrial steroidogenic acute regulatory (StAR) protein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LONP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat